Order Entry
Switzerland
ContactUsLinkComponent
 
Corrosive
Irritant
Trisodium citrate concentrated solution, Sigma-Aldrich®
Catalog # 854C-20ML
CAS Number:  
Corrosive
Irritant
Trisodium citrate concentrated solution, Sigma-Aldrich®
Catalog # 854C-20ML
CAS Number:  

Specifications

  • Pk:
    20 mL
  • Seq:
    2305

Specifications

About this item

 :
 :258.07 g/mol
 :GHS05
 :GHS07
Storage Temperature:Ambient
 :1
CAS Number:68-04-2
Formula:C₆H₅Na₃O₇
MDL Number:MFCD00012462