Antimicrobial foam hand cleanser, OxyBAC®
Katalognr. EVONOXY1LBG
Anbieter: Deb-STOKO
Technische Daten
- Description:Foam hand cleanser with 1 L cartridge
- Pk:6
Technische Daten
Über diesen Artikel
Antimicrobial rich-cream foam hand wash, perfume-free and dye-free.
- Contains accelerated hydrogen peroxide
- Skin hypoallergenic
- Suitable for food facilities and healthcare workers
- Excellent skin cleansing
- Hygienic cartridge
Dispenses foam on the hands which is instantly ready for washing and quick & easy rinsing from hands and sinks. Helps to prevent drying and leave the skin feeling smooth after use.
Certifications: HACCP Certified. Passes EN1499.
Caution: Use biocides safely. Always read the label and product information before use.