Order Entry
Switzerland
ContactUsLinkComponent
VWR® AVS TITRINORM, Cesium chloride/aluminium nitrate buffer solution acc. to schuhknecht and schinkel for determination of na, k and li by flame aas
VWR® AVS TITRINORM, Cesium chloride/aluminium nitrate buffer solution acc. to schuhknecht and schinkel for determination of na, k and li by flame aas
Catalog # 85772.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® AVS TITRINORM, Cesium chloride/aluminium nitrate buffer solution acc. to schuhknecht and schinkel for determination of na, k and li by flame aas
Catalog # 85772.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    Cesium chloride/aluminium nitrate buffer solution

Specifications

About this item

This material is intended for use as an ionisation buffer for AAS

Content of aluminium nitrate nonahydrate (Al(NO₃)₃.9H2O) – 250 g/L
Content of cesium chloride (CsCl) – 50 g/L

Impurities

- K (Potassium) < 1.0 mg/l
- Li (Lithium) < 1.0 mg/l
- Na (Sodium) < 5.0 mg/l