ContactUsLinkComponent
Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Catalog # F4680-25ML
CAS Number:  
Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Catalog # F4680-25ML
CAS Number:  
Specifications
  • Pk:
    25 mL
  • Description:
    Aqueous mounting medium
Specifications
About this item
Fluoromount™ Aqueous Mounting Medium for use with fluorescent dye-stained tissue.
Used for mounting:
- Coverslips on slides for immunofluorescence microscopic examination of PC12 and Hela cells
- Hippocampus neurons cells for confocal microscope studies
- Brain sections for immunofluorescence staining