Orders LinkContactUsLinkComponent
Differentiation solution, Sigma-Aldrich®
Differentiation Solution, Poly bottle, 1 L
  A3179-1L
Differentiation solution, Sigma-Aldrich®
  A3179-1L
 :  
Differentiation solution, Sigma-Aldrich®
Differentiation Solution, Poly bottle, 1 L
 :  
SFr. 57.80
1 * 1 L
Acid Alcohol. Acidified alcohol solution for the differentiation of regressive hematoxylin stains.
  • Suitable for Histology
  • Suitable for Hematology
  • Pack type:
    Poly bottle
  • Pk:
    1 L
  • Description:
    Differentiation Solution
  • Cat. No.:
    A3179-1L
  • M-Clarity quality level:
    MQ200
  • MDL No.:
    MFCD00243254