Order Entry
Switzerland
ContactUsLinkComponent
 
Ammonium chloride for molecular biology
Catalog # ACRO464595000
CAS Number:  
undefined
Ammonium chloride for molecular biology
Catalog # ACRO464595000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder, crystals and/or chunks
  • Solubility:
    (10 % in water) Clear colorless solution
  • Titration Argentimetric:
    ≥99.4 %
  • pH:
    4.5 to 5.5
  • Dnase,Rnase,Protease activity:
    none detected
  • Phosphate (PO4):
    ≤2 ppm
  • Sulfate (SO4):
    ≤20 ppm
  • Iron (Fe):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

About this item

 :
 :53.49 g/mol
Melting Pt:340 °C (sublimes) (subl.)
 :GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420
Melting Pt:340 °C (subl.)