Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Chloroform-[D₁] 99.8 atom % D + 0.1 v/v% TMS stabilised for NMR spectroscopy
  ACRO464430250
 :  
undefined
 
 
Chloroform-[D₁] 99.8 atom % D + 0.1 v/v% TMS stabilised for NMR spectroscopy
  ACRO464430250
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 %
  • D-Enrichment:
    ≥99.75 %
  • TMS:
    0.09 to 0.11 v/v %

 

 

 :
 :120.38 g/mol
 :60 °C (760 mmHg)
 :–64 °C
 :Ambient
 :6.1,III
 :Stabilised with silver
 :1888
 :GHS06
 :GHS08
 :865-49-6
 :CDCl₃
 :MFCD00000827