

Ethanol denatured ≥96%, extra pure (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78 °C (760 mmHg)
Melting Pt:–114.1 °C
Density:0.79
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1170
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.3620 to 1.3640 (20°C, 589 nm)
- Density:(20°C) 0.803 to 0.812 kg/l
- Additional info:Denatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex
- Ethanol:95 to 97 % v/v (calculated)
Specification Test Results
Frequently Bought Together