Order Entry
Switzerland
ContactUsLinkComponent
Ethanol denatured (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Flammable
Irritant
Ethanol denatured (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Certificates
undefined
Flammable
Irritant
Ethanol denatured (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.3610 to 1.3630 (20°C, 589 nm)
  • Water:
    ≤3400 ppm
  • Density:
    (20°C) 0.789 to 0.790 kg/l
  • Additional info:
    Denatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex
  • Ethanol:
    99.8 to 100.0 % v/v (calculated)

Specification Test Results

Frequently Bought Together