Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Fast HiFi DNA polymerase, 2X master mix
VWR®, Fast HiFi DNA polymerase, 2X master mix
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Fast HiFi DNA polymerase, 2X master mix
Supplier:  VWR Chemicals

Specifications

  • Application:
    Cloning (high fidelity)
    Standard amplification
  • Format:
    Mastermix incl. dNTPs
  • High fidelity:
    Yes
  • Hot start:
    No
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Amplicon size:
    11 kb for gDNA
  • Processivity:
    Fast
  • Exonuclease activity:
    5’→3’ 3’→5’
  • Product overhang:
    No
  • Features:
    feature-ls-33964141|feature-ls-33964143
  • Concentration:
    2x
  • Description:
    Fast HiFi DNA Polymerase 2x Master Mix 500 reactions
  • Size:
    10 x 1.25 ml
  • Units:
    500 r Mix 500

Specifications

Frequently Bought Together