ContactUsLinkComponent
Ladles, long handle, detectable, SteriPlast®
Ladles, long handle, detectable, SteriPlast®
Catalog # 331-0361
Supplier:  BURKLE
undefined
Ladles, long handle, detectable, SteriPlast®
Catalog # 331-0361
Supplier:  BURKLE
Specifications
  • Sterile:
    +
  • Capacity:
    30 ml
  • Colour:
    Blue
  • Pk:
    10
Specifications
About this item
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work.
  • PS detectable
  • Individually packaged and sterilised, for single use
  • Reaches into every corner, handle length 300 mm
  • Can be detected by colour scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
The disposable ladle can be used in the pharmaceutical industry, chemistry and cosmetics and the food and animal feed industries.