ContactUsLinkComponent
Kimtech Science™ Kimwipes™ Wipes for Delicate Tasks
Kimtech Science™ Kimwipes™ Wipes for Delicate Tasks
Catalog # 115-0284
Supplier:  KIMBERLY CLARK
undefined
Kimtech Science™ Kimwipes™ Wipes for Delicate Tasks
Catalog # 115-0284
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    60 boxes of 280 sheets
  • Colour:
    White
  • Description:
    Delicate task wipes, 1 ply tissue
  • L×W:
    112×213 mm
  • Seq:
    03
  • Pk:
    280
Specifications
About this item
These light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.
  • Anti-stat dispensing reduces lint and electrostatic discharge and controls usage
  • Portability reduces waste
  • One-at-a-time dispensing (Pop-Up™ Box dispenser)
  • Also available in 2-ply and 3-ply for increased absorbency
  • Gentle texture
Used in environments like research facilities, pharmaceutical, biotech, high-tech, manufacturing.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904