ContactUsLinkComponent
Chromate test strips, Dosatest
Chromate test strips, Dosatest
  85695.601
 :  
undefined
Chromate test strips, Dosatest
  85695.601
 :  
  • Chemical method:
    Carbazide
  • Pk:
    100
  • Description:
    Chromate test strips
  • Range:
    0 • 3 • 10 • 30 • 100  mg/L CrO₄²⁻
Semi-quantitative test strips for the determination of chromate.
  • Ideal for rapid and easy analysis of chromate directly on-site
  • No maintenance or calibration is required
Colour change: White to violet.
 : Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box