Sterilisation indicating labels for thermal transfer, series R4300
Supplier: Brady
Certificates
About this item
This B-7425-AC material is a steam sterilisation indicating polypropylene label with a preprinted header that displays 'STERILIZED' when exposed to autoclave conditions. It's ideal for use in sample tracking, inventory management, general tracking, and identification of laboratory materials that require sterilisation.
- Die-cut labels are on a roll
- Application: general identification, laboratory labelling, temperature indicating labelling
- This R4300 label has a matt finish
- Service temperature: –80 to 70 °C
- Thickness: 0,102 mm
- Autoclavable and cryogenic storage are the material's properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.
This economical solution for identifying vials, tubes, racks and flasks adheres to stainless steel, glass and polypropylene, and can withstand temperatures from –196 to 121°C.