Auftragserfassung
Switzerland
ContactUsLinkComponent
 
Arsenic test strips, Dosatest
Catalog # 85710.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Arsenic test strips, Dosatest
Catalog # 85710.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemische Methode:
    Gutzeit (modified)
  • VE:
    100
  • Bezeichnung:
    Arsenic test strips
  • Tests pro Kit:
    100
  • Messbereich:
    0 • 0.01 • 0.025 • 0.05 • 0.1 • 0.5  mg/L As³⁺ ⁵⁺

Specifications

About this item

Semi-quantitative test strips for the determination of arsenic.

  • Ideal for rapid and easy analysis of arsenic directly on-site
  • No maintenance or calibration is required

Colour change: White to yellow-brown

Delivery: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box
Caution: Each test field on the test strips contains mercury(II) bromide. Toxic in contact with skin