Order Entry
Switzerland
ContactUsLinkComponent
Ammonium Test Strips, Dosatest
Ammonium Test Strips, Dosatest
Catalog # 85702.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium Test Strips, Dosatest
Catalog # 85702.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Nesslers reagent
  • Pk:
    100
  • Description:
    Ammonium test strips
  • Tests per kit:
    100
  • Range:
    0 • 10 • 25 • 50 • 100 • 200 • 400  mg/L NH₄⁺

Specifications

About this item

Semi-quantitative test strips for the determination of ammonium.

  • Ideal for rapid and easy analysis of ammonium directly on-site
  • No maintenance or calibration is required

Colour change: Bright yellow to orange.

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.