Order Entry
Switzerland
ContactUsLinkComponent
Bovine MSG-Trypsin™
Bovine MSG-Trypsin™
Supplier:  G-Biosciences
Certificates
undefined
Bovine MSG-Trypsin™
Supplier:  G-Biosciences

Specifications

  • Species:
    Bovine
  • Enzyme name:
    MSG-Trypsin™
  • Grade:
    Mass Spectroscopy Grade

Specifications

Related Information

Frequently Bought Together