Examination gloves, nitrile, Gen-X®, smart glove
Supplier: SMART GLOVE
Certificates
About this item
These non sterile examination-grade gloves are made of nitrile.
- Ambidextrous
- Fully palm textured to provide a secure grip
- Extra-soft and comfortable
- Powder-free
- Beaded cuffs allow easy donning
- 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
Ideal for use in industrial and laboratory applications.
Certifications: EN ISO 374-1, EN 374-2, EN 374-4, EN374-5, EN 16523-1; Cat. III; AQL 1,5
Packaging: 100 gloves per box.
Specifications
- Catalog No:
- 112-6136
- 112-6133
- 112-6137
- 112-6134
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Palm textured
- Palm textured
- Palm textured
- Palm textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded