Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Vanadium(V) oxide ≥98%, GPR RECTAPUR®

Health
Irritant
Environment
Supplier:   VWR Chemicals
Vanadium(V) oxide ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Vanadium(V) oxide ≥98%, GPR RECTAPUR®

2 Options Available Below

Product Details & Documents
Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • UN:2862
  • ADR:6.1,III
Specifications for Product Family

Specification Test Results

  • Assay:
    ≥ 98 %
  • Appearance:
    Brown crystalline powder
  • Total N (Nitrogen):
    ≤ 0.1 %
  • Cl (Chloride):
    ≤ 0.1 %
  • PO₄ (Phosphate):
    ≤ 0.1 %
  • SO₄ (Sulphate):
    ≤ 0.1 %
  • Fe (Iron):
    ≤ 0.1 %
Product Family Options

Product Information