ContactUsLinkComponent
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
Catalog # 215-4281
Supplier:  DWK Life Sciences
   Waste Reducing
undefined
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
Catalog # 215-4281
Supplier:  DWK Life Sciences
   Waste Reducing
Specifications
  • Description:
    High-temperature cap liner, GL 45, PTFE coated FKM, 43,5×3,0 mm
  • Environmentally Preferable:
    Waste Reducing
  • Pk:
    10
Specifications
About this item
Replacement GL 45 PTFE coated high-temperature silicone cap liner for the aluminium cap.