Professional refrigerators, SRR, PHCbi
Anbieter: PHC Corporation
Zertifikate
Über diesen Artikel
Designed for general purpose storage applications in clinical, life science, pharmaceutical, biotechnology, and industrial laboratories.
- Microprocessor temperature control with LED readout and alarm functions
- Large interiors with greater flexibility in cold storage
- Reliable heavy duty refrigeration systems for frequent door openings
- Forced air circulation refrigeration system with automatic evaporator defrost; Automatic condensate evaporator
- Stainless steel exterior (430 series) and interior (304 series) for long life and durability
- Automatic interior lighting, 5-inch casters, door locks
- Heavy duty epoxy coated shelves per section mounted on stainless steel pilasters
- Hinged panel allows for easy servicing
- Environmentally friendly R134A refrigerant
SRR-23FD-MED & SRR-49FD-MED: Solid swing-door refrigerators
SRR-23GD-MED, SRR-49GD-MED, SRR-72GD-MED: Glass swing-door refrigerators
Same features as above plus:
Energy efficient triple pane glass doors, without door lock
Temperature range: −3 to +10 °C
Same features as above plus:
Frequently Bought Together