ContactUsLinkComponent
Cleanroom gloves, polychloroprene, sterile, BioClean Ultimate™
 :  Nitritex
undefined
Cleanroom gloves, polychloroprene, sterile, BioClean Ultimate™
 :  Nitritex
  • Catalog No:
  • 112-1660
  • 112-1666
  • 112-1662
  • 112-1661
  • 112-1664
  • 112-1665
  • 112-1663
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • Finger thickness:
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • Palm thickness:
  • 0,11 mm
  • 0,11 mm
  • 0,11 mm
  • 0,11 mm
  • 0,11 mm
  • 0,11 mm
  • 0,11 mm
  • EN norm:
  • EN 374, EN 455
  • EN 374, EN 455
  • EN 374, EN 455
  • EN 374, EN 455
  • EN 374, EN 455
  • EN 374, EN 455
  • EN 374, EN 455
Frequently Bought Together