Order Entry
Switzerland
ContactUsLinkComponent
pH/mV/°C/ISE meter, bench, Orion™ Dual Star™
pH/mV/°C/ISE meter, bench, Orion™ Dual Star™
Supplier:  Thermo Orion
Certificates
undefined
pH/mV/°C/ISE meter, bench, Orion™ Dual Star™
Supplier:  Thermo Orion

Specifications

  • Catalog No:
  • ORIO2115203
  • ORIO2115001
  • ORIO2115101
  • ORIO2115205
  • ORIO2115000
  • ORIO2115102
  • ORIO2115201
  • ORIO2115204
  • ORIO2115202
  • Model:
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • Dual Star™
  • pH range:
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • −2,000...+19,999
  • pH resolution:
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • 0,1, 0,01, 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • 1 - 6 points with automatic buffer recognition (NIST, DIN, custom)
  • mV range:
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • ±1999,9
  • mV resolution:
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ±0,2 of measured value or 0,05% of reading (whichever is greater)
  • ±0,2 of measured value or 0,05% of reading (whichever is greater)
  • ±0,2 of measured value or 0,05% of reading (whichever is greater)
  • ±0,2 of measured value or 0,05% of reading (whichever is greater)
  • ±0,2 of measured value or 0,05% of reading (whichever is greater)
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ISE range:
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • 0 - 19999
  • ISE resolution:
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • ISE accuracy:
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ±0,2 of measured value or 0,05%
  • ±0,2 of measured value or 0,05%
  • ±0,2 of measured value or 0,05%
  • ±0,2 of measured value or 0,05%
  • ±0,2 of measured value or 0,05%
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ±0,2 mV or ±0,05% of reading; whichever is greater
  • ISE calibration:
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • 2 - 6 points (linear, point to point, non-linear selectable auto-blank)
  • ISE units:
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • ppm, ppb, M, % or unitless
  • Temperature range:
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • –5...+105 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Temperature calibration:
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • Temperature compensation:
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Power supply:
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Universal AC adapter (included)
  • Compliance:
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • Operating temperature range:
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • 5…45 °C
  • Data logging - log function:
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Automatic data logging with auto-read, on ready, at timed intervals, value change in measurement and timed reading measure modes; manual data logging with continuous measure mode
  • Data logging capacity:
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • 1000 points with time/date stamp
  • Memory:
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • 20 methods (10 per channel with calibration data)
  • Languages:
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • English, Spanish, French, Italian, and German
  • IP rating:
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • Dimensions:
  • 24L×17W×9H cm (9,5×6,7×3,5")
  • 24L×17W×9H cm (9.5×6.7×3.5")
  • 24L×17W×9H cm (9.5×6.7×3.5")
  • 24L×17W×9H cm (9,5×6,7×3,5")
  • 24L×17W×9H cm (9.5×6.7×3.5")
  • 24L×17W×9H cm (9.5×6.7×3.5")
  • 24L×17W×9H cm (9,5×6,7×3,5")
  • 24L×17W×9H cm (9,5×6,7×3,5")
  • 24L×17W×9H cm (9,5×6,7×3,5")
  • Connections:
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Outputs:
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • USB, RS232
  • Input:
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Alarm:
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • High/low limit alarm
  • Parameters:
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
  • Calibration:
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ.
  • Calibration points:
  • 1 point mV (RmV) mode; 1 point ORP mode; 2 - 6 points ISE mode; 1 - 6 points pH mode
  • 1 point mV (RmV) mode; 1 point ORP mode; 2 - 6 points ISE mode; 1 - 6 points pH mode
  • 1 point mV (RmV) mode; 1 point ORP mode; 2 - 6 points ISE mode; 1 - 6 points pH mode
  • 1 point mV (RmV) mode; 1 point ORP mode; 2 - 6 points ISE mode; 1 - 6 points pH mode
  • W×D×H:
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • 240×170×90 mm
  • Weight:
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg

Specifications

Electrodes

Product Information

Frequently Bought Together