ContactUsLinkComponent
Pestle for micro-tissue grinders with screw caps
Pestle for micro-tissue grinders with screw caps
Catalog # 432-5004
Supplier:  DWK Life Sciences
undefined
Pestle for micro-tissue grinders with screw caps
Catalog # 432-5004
Supplier:  DWK Life Sciences
Specifications
  • For:
    Wheaton micro-tissue grinder with screw caps
  • Description:
    Pestle
  • Pk:
    2
Specifications
About this item
Replacement pestle for WHEATON® micro tissue grinder with cap, nominal capacity 0,5 ml.
  • Manufactured from borosilicate glass 3.3
  • With PTFE pestle and screw cap
  • 13-425 open top screw cap and PTFE faced silicone rubber liner
  • Autoclavable