VWR®, Turbidity standards, formazin
Fournisseur: VWR Chemicals
Certificats
À propos de cet article
All turbidity standard solutions are Certified Reference Materials.
This standard also meets the requirements of the European Pharmacopoeia on a “2.2.1. Clarity and degree of opalescence of liquids” and ISO 7027-1 “Water quality – Determination of turbidity – part 1: Quantitative methods".
Certifications: Produced and calibrated under a quality system that is:
- ISO 9001 certified
- Accredited according to ISO/IEC 17025 - Testing
- Accredited according to ISO 17034 - Reference Material Producer
- ISO 9001 certified
- Accredited according to ISO/IEC 17025 - Testing
- Accredited according to ISO 17034 - Reference Material Producer
Informations associées
Certificats
Rechercher par numéro de lotSDS_EN_CH_VWRC88232.pdf SDS_EN_CH_VWRC88233.pdf SDS_EN_CH_VWRC88235.pdf SDS_EN_CH_VWRC88236.pdf SDS_EN_CH_VWRC88237.pdf
FDS
Plus
VWR® for Calibrations
Documentation produit
Frequently Bought Together