Avantor® Hichrom UNCORET, GC Columns
Anbieter: Avantor
Zertifikate
Über diesen Artikel
Large volume injection pre-columns.
- Excellent reproducibility
- High separation efficiency
Technische Daten
- Catalog No:
- HIU-53-045-12G3
- HIU-53-025-12G3
- Phase name:
- UNCORET
- UNCORET
- Phase composition:
- 5% Phenyl 95% Methyl Polysiloxane
- 5% Phenyl 95% Methyl Polysiloxane
- Polarity:
- Polar stationary phase
- Polar stationary phase
- Crossbond:
- Yes
- Yes
Technische Daten
Frequently Bought Together