SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Analytical standard, Gibberellic acid, reference material, Fluka™
Analytical standard, Gibberellic acid, reference material, Fluka™
HONCR1768-100MG
:
Honeywell Chemicals
:
Analytical standard, Gibberellic acid, reference material, Fluka™
HONCR1768-100MG
:
Honeywell Chemicals
:
Pack type:
Glass bottle
Pk:
100 mg
Standards:
Reference material
CAS No.:
77-06-5
Description:
Gibberellic acid
Description:
Gibberellic acid
Form:
Neat