Order Entry
Switzerland
ContactUsLinkComponent
 
Analytical standard, Imazethapyr, reference material, Fluka™
  HONCR1731-100MG
 :  
Analytical standard, Imazethapyr, reference material, Fluka™
  HONCR1731-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    81335-77-5
  • Description:
    Imazethapyr
  • Description:
    Imazethapyr
  • Form:
    Neat