Analytical standard, Dimethomorph (mixture of E+Z isomers), reference material, Fluka™
HONCR1791-100MG
:
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:110488-70-5
- Description:Dimethomorph (mixture of E+Z isomers)
- Description:Dimethomorph (mixture of E+Z isomers)
- Form:Neat