Analytical standard, Ampicillin-d₅, reference material, Fluka™
Catalog # HONCR1758-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Standards:Reference material
- CAS No.:1426173-65-0
- Description:Ampicillin-d5
- Formula:C₁₆H₁₄D₅N₃O₄S
- Storage temperature:2…8 °C
- Description:Ampicillin-d5
- Form:Neat