Calcium AAS standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Frequently Bought Together