Triethylamine Standard for Ion Chromatography, (CRM), Fluka™
Codice catalogo HONCE3053-100ML
Nome del fornitore: Honeywell Chemicals
Numero CAS:
Specifiche
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Triethylamine standard
Specifiche
Informazioni su questo articolo
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).