Order Entry
Switzerland
ContactUsLinkComponent
Nitrite (N) standard for ion chromatography, (CRM), Fluka™
Nitrite (N) standard for ion chromatography, (CRM), Fluka™
Catalog # HONCE3035-100ML
CAS Number:  
Nitrite (N) standard for ion chromatography, (CRM), Fluka™
Catalog # HONCE3035-100ML
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Nitrite (N) standard for IC

Specifications

About this item

Certified Reference Material (CRM)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).