Order Entry
Switzerland
ContactUsLinkComponent
Selenium ICP & ICP-MS Standards, (CRMs), Fluka™
Selenium ICP & ICP-MS Standards, (CRMs), Fluka™
Catalog # HONCE1418-100ML
CAS Number:  
Selenium ICP & ICP-MS Standards, (CRMs), Fluka™
Catalog # HONCE1418-100ML
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).