Order Entry
Switzerland
ContactUsLinkComponent
Antimony ICP & ICP-MS Standards, (CRMs), Fluka™
Antimony ICP & ICP-MS Standards, (CRMs), Fluka™
  HONCE1416-100ML
 :  
Antimony ICP & ICP-MS Standards, (CRMs), Fluka™
  HONCE1416-100ML
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃/0,5% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Antimony
  • Symbol:
    Sb

 

 

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
 : ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).