Order Entry
Switzerland
ContactUsLinkComponent
 
Quality control standard 3 for ICP
Catalog # HONCE2212-100ML
CAS Number:  
Quality control standard 3 for ICP
Catalog # HONCE2212-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Quality Control Standard 3

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Al, Ba, Cd, Ca, Cr, Co, Cu, Fe, Pb, Mg, Mn, Ni, Na, Ti, Zn in HNO₃ 5%