NexION standard/DRC mode detection limit solution
Catalog # HONCE2192-100ML
Anbieter: Honeywell Chemicals
CAS-Nummer:
Technische Daten
- Matrix:0,5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Description:NexION Standard/DRC Mode Detection Limit Solution
Technische Daten
Über diesen Artikel
Certified Reference Material (CRM).
Blank Solution in HNO₃ 0.5%