Order Entry
Switzerland
ContactUsLinkComponent
SmartTune solution for standard ELAN/DRC-e Hg for ICP
SmartTune solution for standard ELAN/DRC-e Hg for ICP
Catalog # HONCE2186-100ML
CAS Number:  
SmartTune solution for standard ELAN/DRC-e Hg for ICP
Catalog # HONCE2186-100ML
CAS Number:  

Specifications

  • Matrix:
    1% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    SmartTune Solution

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Ba, Be, Ce, Co, In, Mg, Pb, Rh, U in HNO₃ 1%