Order Entry
Switzerland
ContactUsLinkComponent
Elan 6000/5000 plasma setup solution Hg for ICP
Elan 6000/5000 plasma setup solution Hg for ICP
Catalog # HONCE2185-100ML
CAS Number:  
Elan 6000/5000 plasma setup solution Hg for ICP
Catalog # HONCE2185-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Elan 6000/5000 Plasma Setup Solution Hg

Specifications

About this item

Certified Reference Material (CRM).

10 g/L each of Ba, Cd, Ce, Cu, Ge, Pb, Mg, Rh, Sc, Tb, Tl in HNO₃ 2%