ContactUsLinkComponent
Trace Metals III for AA and ICP-OES
  HONCE2168-100ML
 :  
Trace Metals III for AA and ICP-OES
  HONCE2168-100ML
 :  
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Trace Metals III
Certified Reference Material (CRM).
500 mg/L each of Ba, Ca, Mo, Na, 100 mg/L each of K, Mg in HNO₃ 2%