Order Entry
Switzerland
ContactUsLinkComponent
Selenium Solution for AA and ICP-OES, Fluka™
Selenium Solution for AA and ICP-OES, Fluka™
Catalog # HONCE2161-100ML
CAS Number:  
Selenium Solution for AA and ICP-OES, Fluka™
Catalog # HONCE2161-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Selenium Solution

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L Selenium (Se) in HNO₃ 5%