ContactUsLinkComponent
TCLP Standard 1 for ICP
  HONCE2160-100ML
 :  
TCLP Standard 1 for ICP
  HONCE2160-100ML
 :  
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    TCLP Standard 1
Certified Reference Material (CRM).
500 mg/L Ba, 100 mg/L Hg, 25 mg/L each of Ag, As, Cr, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%