Order Entry
Switzerland
ContactUsLinkComponent
MS environmental standard for ICP-MS
MS environmental standard for ICP-MS
Catalog # HONCE2152-100ML
CAS Number:  
MS environmental standard for ICP-MS
Catalog # HONCE2152-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    MS Environmental Standard

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn in HNO₃ 5%