Order Entry
Switzerland
ContactUsLinkComponent
Instrument calibration standard 3 for ICP
Instrument calibration standard 3 for ICP
Catalog # HONCE2150-100ML
CAS Number:  
Instrument calibration standard 3 for ICP
Catalog # HONCE2150-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 3

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L each of Ca, Fe, K, Mg, Na in HNO₃ 5%