Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Multi-element solution for ICP, (17 Components)

Multi-element solution for ICP, (17 Components)

Multi-Element Solution for ICP, (17 components), 5% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2134-100ML
Supplier:   Honeywell Chemicals
Multi-element solution for ICP, (17 Components)
Catalog # HONCE2134-100ML
CAS Number:  
Multi-element solution for ICP, (17 Components)

Multi-Element Solution for ICP, (17 components), 5% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
SFr. 944.93
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

1000 mg/L each of As, Ba, Cd, Be, Cr, Co, Cu, La, Pb, Li, Mn, Ni, Sc, Sr, V, Y, Zn in HNO₃ 5%

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Solution for ICP, (17 components)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.