Order Entry
Switzerland
ContactUsLinkComponent
Spike sample standard water for ICP
Spike sample standard water for ICP
Catalog # HONCE2128-100ML
CAS Number:  
Spike sample standard water for ICP
Catalog # HONCE2128-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Spike Sample Standard Water

Specifications

About this item

Certified Reference Material (CRM).

500 mg/L Fe, 250 mg/L each of Ba, Zn, 100 mg/L each of Co, Cr, Cu, Mn, Ni, Sb, V, 50 mg/L each of As, Pb, 25 mg/L each of Ag, Be, Cd, Se, Tl in HNO₃ 5%