Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Interference check solution 2 for ICP

Interference check solution 2 for ICP

Interference Check Solution 2, 2% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2119-100ML
Supplier:   Honeywell Chemicals
Interference check solution 2 for ICP
Catalog # HONCE2119-100ML
CAS Number:  
Interference check solution 2 for ICP

Interference Check Solution 2, 2% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
SFr. 480.76
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

10 mg/L each of Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn in HNO₃ 2%

Specifications

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference Check Solution 2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.