Order Entry
Switzerland
ContactUsLinkComponent
 
Instrument check standard 4 for ICP
Catalog # HONCE2110-100ML
CAS Number:  
Instrument check standard 4 for ICP
Catalog # HONCE2110-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Check Standard 4

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Mo, Th, U in HNO₃ 2%