Order Entry
Switzerland
ContactUsLinkComponent
Instrument Calibration standard 4 for ICP and HGA elements
Instrument Calibration standard 4 for ICP and HGA elements
Catalog # HONCE2106-100ML
CAS Number:  
Instrument Calibration standard 4 for ICP and HGA elements
Catalog # HONCE2106-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 4

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of As, Tl, 50 mg/L each of Cd, Se, 30 mg/L Pb in HNO₃ 5%