Order Entry
Switzerland
ContactUsLinkComponent
 
Quality control standard pure XVI for ICP
Catalog # HONCE2101-100ML
CAS Number:  
Quality control standard pure XVI for ICP
Catalog # HONCE2101-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Quality Control Standard Pure XVI

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in HNO₃ 5%